Methyl carbazate ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Methyl hydrazinecarboxylate, Methyl hydrazinocarboxylate
A14548.0E
A14548.22
A14548.0EEA
21520
CZK
A14548.0E
A14548.22
Methyl carbazate ≥98%
Methyl carbazate
Purity: 98%
CAS No.: 6294-89-9
Molecular Formula: C2H6N2O2
Formula Weight: 90.08
Odour: Weak
Melting Point: 70-73°
Beilstein Reference: 1679395
UN No.: 2811
MDL No.: MFCD00007594
Vzorec:
C₂H₆N₂O₂ MW: 90,08 g/mol Bod tání: 70…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007594 CAS číslo: 6294-89-9 EINECS: 228-560-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...