Ethyl carbazate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13860.0B
A13860.18
A13860.30
A13860.0BEA
19360
CZK
A13860.0B
A13860.18
A13860.30
Ethyl carbazate ≥97%
Ethyl carbazate
Purity: 97%
CAS No.: 4114-31-2
Molecular Formula: C3H8N2O2
Formula Weight: 104.11
Flash Point: 86°(186°F)
Melting Point: 44-47°
Boiling Point: 85-86°/7mm
Beilstein Reference: 878265
UN No.: 2811
MDL No.: MFCD00007595
Vzorec:
C₃H₈N₂O₂ Bod varu: 85…86 °C (7 mmHg) Bod tání: 44…47 °C Bod vzplanutí: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007595 CAS číslo: 4114-31-2 EINECS: 223-903-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...