Tisk…

tert-Butyl carbazate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: tert-Butyl hydrazinecarboxylate

M-Clarity™ quality level = MQ200

B91005-100G B91005-25G B91005-500G B91005-5G
SIALB91005-100GEA 8390 CZK
SIALB91005-100G SIALB91005-25G SIALB91005-500G SIALB91005-5G
tert-Butyl carbazate, Sigma-Aldrich®
tert-Butyl carbazate
Vzorec: C₅H₁₂N₂O₂
MW: 132,16 g/mol
MDL Number: MFCD00007593
CAS číslo: 870-46-2
EINECS: 212-795-3
UN: 1325
ADR: 4.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR