tert-Butyl carbazate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
tert-Butyl hydrazinecarboxylate
A12383.14
A12383.22
A12383.36
A12383.14EA
2330
CZK
A12383.14
A12383.22
A12383.36
tert-Butyl carbazate ≥98%
tert-Butyl carbazate
Purity: 98+%
CAS No.: 870-46-2
Molecular Formula: C5H12N2O2
Formula Weight: 132.16
Storage Temperature: 2°C to 8°C
Flash Point: 91°(195°F)
Melting Point: 37-41°
Boiling Point: 63-65°/0.1mm
Beilstein Reference: 1756967
UN No.: 1325
MDL No.: MFCD00007593
Vzorec:
C₅H₁₂N₂O₂ Bod varu: 63…65 °C (0,1 mmHg) Bod tání: 37…41 °C Bod vzplanutí: 91 °C (195 °F) |
MDL Number:
MFCD00007593 CAS číslo: 870-46-2 EINECS: 212-795-3 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...