Tisk…

Trifluoromethanesulphonic anhydride 1 mol/l in dichloromethane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Triflic anhydride

M-Clarity™ quality level = MQ100

704083-100ML 704083-25ML
SIAL704083-100MLEA 8240 CZK
SIAL704083-100ML SIAL704083-25ML
Trifluoromethanesulphonic anhydride 1 mol/l in dichloromethane, Sigma-Aldrich®
Trifluoromethanesulphonic anhydride
Vzorec: C₂F₆O₅S₂
MW: 282,14 g/mol
MDL Number: MFCD00000408
CAS číslo: 358-23-6
EINECS: 206-616-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR