(±)-2-Bromopropionyl bromide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
249785-100G
249785-500G
SIAL249785-100GEA
1400
CZK
SIAL249785-100G
SIAL249785-500G
(±)-2-Bromopropionyl bromide, Sigma-Aldrich®
(±)-2-Bromopropionyl bromide
Vzorec:
C₃H₄Br₂O MW: 215,87 g/mol |
MDL Number:
MFCD00000142 CAS číslo: 563-76-8 EINECS: 209-261-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...