Tisk…

6-Methoxy-2-methylbenzothiazole ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Methoxy-2-methylbenzo[d]thiazole

H32425.03 H32425.06
H32425.03EA 2040 CZK
H32425.03 H32425.06
6-Methoxy-2-methylbenzothiazole ≥97%
6-Methoxy-2-methylbenzothiazole

Purity: 97% CAS No.: 2941-72-2 Molecular Formula: C9H9NOS Formula Weight: 179.24 Flash Point: 109°(228°F)

Vzorec: C₉H₉NOS
MW: 179,24 g/mol
Hustota: 1,204
Bod vzplanutí: 109 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005795
CAS číslo: 2941-72-2
EINECS: 220-931-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR