Tisk…

Diisoamyl ether (mixture of isomers), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

260649-100ML 260649-500ML
SIAL260649-100MLEA 1400 CZK
SIAL260649-100ML SIAL260649-500ML
Diisoamyl ether (mixture of isomers), Sigma-Aldrich®
Diisoamyl ether (mixture of isomers)
Vzorec: C₁₀H₂₂O
MW: 158,28 g/mol
MDL Number: MFCD00008947
CAS číslo: 544-01-4
EINECS: 208-857-4
UN: 3271
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR