Tisk…

Sesamol-(3,4-(methylenedioxy)phenol), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sesamol, 3,4-Methylenedioxyphenol, Benzo[d][1,3]dioxol-5-ol

M-Clarity™ quality level = MQ100

S3003-25G-A S3003-5G-A
SIALS3003-25G-AEA 2670 CZK
SIALS3003-25G-A SIALS3003-5G-A
Sesamol-(3,4-(methylenedioxy)phenol), Sigma-Aldrich®
Sesamol-(3,4-(methylenedioxy)phenol)
Vzorec: C₇H₆O₃
MW: 138.12
MDL Number: MFCD00005827
CAS číslo: 533-31-3
EINECS: 208-561-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR