3,4-(Methylenedioxy)aniline ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
5-Amino-1,3-benzodioxole
A12975.09
A12975.18
A12975.30
A12975.09EA
968
CZK
A12975.09
A12975.18
A12975.30
3,4-(Methylenedioxy)aniline ≥98%
3,4-(Methylenedioxy)aniline
Purity: 98+%
CAS No.: 14268-66-7
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 142°(287°F)
Melting Point: 40-45°
Boiling Point: 144°/16mm
Beilstein Reference: 4919
UN No.: 2811
MDL No.: MFCD00005832
Vzorec:
C₇H₇NO₂ Bod varu: 144 °C (16 mmHg) Bod tání: 40…45 °C Bod vzplanutí: 142 °C (287 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005832 CAS číslo: 14268-66-7 EINECS: 238-161-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...