2-Imidazolidinethione for synthesis, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
8.20696.0250
8.20696.1000
8.20696.0250EA
1750
CZK
8.20696.0250
8.20696.1000
2-Imidazolidinethione for synthesis, Sigma-Aldrich®
2-Imidazolidinethione
Vzorec:
C₃H₆N₂S Storage Temperature: Ambient |
MDL Number:
MFCD00005276 CAS číslo: 96-45-7 EINECS: 202-506-9 |
Výsledky specifikačního testu
Solubility | 8 g/l (20 °C) |
Melting Point | 196 - 199 °C |
Molar Mass | 102.16 g/mol |
Bulk Density | 750 - 900 kg/m3 |
Vapor Pressure | 0.2 hPa (170 °C) |
Flash Point | 253 °C |
Density | 1.4 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...