Tisk…

(±)-Ethyl-2,3-dibromopropionate ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15591.14 A15591.22 A15591.36
A15591.14EA 927 CZK
A15591.14 A15591.22 A15591.36
(±)-Ethyl-2,3-dibromopropionate ≥97%
(±)-Ethyl-2,3-dibromopropionate

Purity: 97% CAS No.: 3674-13-3 Molecular Formula: C5H8Br2O2 Formula Weight: 259.94 Flash Point: 90°(194°F) Refractive Index: 1.5 00 Odour: Irritating Boiling Point: 212-214° Beilstein Reference: 636046 UN No.: 2922 MDL No.: MFCD00000212

Vzorec: C₅H₈Br₂O₂
Bod varu: 212…214 °C
Hustota: 1,789
Bod vzplanutí: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000212
CAS číslo: 3674-13-3
EINECS: 222-941-8
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR