1-Amino-4-methylpiperazine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Methylpiperazin-1-amine
L19108.22
L19108.14
L19108.22EA
3350
CZK
L19108.22
L19108.14
1-Amino-4-methylpiperazine 98%
1-Amino-4-methylpiperazine
Purity: 98%
CAS No.: 6928-85-4
Molecular Formula: C5H13N3
Formula Weight: 115.18
Flash Point: 70°(158°F)
Refractive Index: 1.4840
Boiling Point: 172-175°
Beilstein Reference: 103334
UN No.: 2735
MDL No.: MFCD00006154
Vzorec:
C₅H₁₃N₃ Bod varu: 172…175 °C Hustota: 0,960 Bod vzplanutí: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006154 CAS číslo: 6928-85-4 EINECS: 230-053-7 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...