Tisk…

Isobutyl propionate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ300

W221201-1KG-K W221201-4KG-K
SIALW221201-1KG-KEA 4070 CZK
SIALW221201-1KG-K SIALW221201-4KG-K
Isobutyl propionate, Sigma-Aldrich®
Isobutyl propionate
Vzorec: C₇H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00009307
CAS číslo: 540-42-1
EINECS: 208-746-0
UN: 1914
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR