Tisk…

Dimethoxy(diisobutyl)silane for synthesis, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diisobutyldimethoxysilane

M-Clarity™ quality level = MQ200

8.14720.0025 8.14720.0100
8.14720.0025EA 2010 CZK
8.14720.0025 8.14720.0100
Dimethoxy(diisobutyl)silane for synthesis, Sigma-Aldrich®
Dimethoxy(diisobutyl)silane
Vzorec: C₁₀H₂₄O₂Si
MW: 204,38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236510
CAS číslo: 17980-32-4
UN: 3082
ADR: 9,III

Objednat

Výsledky specifikačního testu

Solubility 13.4 mg/l (25 °C) Hydrolysis
Molar Mass 204.38 g/mol
Boiling Point 188 °C (1025 hPa)
Vapor Pressure 0.75 hPa (25 °C)
Flash Point 62 °C
Density 0.863 g/cm3 (20 °C)
Ignition Point 255 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR