Ethyl propionylacetate 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl 3-oxopentanoate, Ethyl 3-oxovalerate
A15464.06
A15464.14
A15464.22
A15464.06EA
2230
CZK
A15464.06
A15464.14
A15464.22
Ethyl propionylacetate 95%
Ethyl propionylacetate
Purity: 95%
CAS No.: 4949-44-4
Molecular Formula: CH3CH2COCH2CO2CH2CH3
Formula Weight: 144.17
Flash Point: 77°(170°F)
Refractive Index: 1.4220
Boiling Point: 190-192°
Beilstein Reference: 970270
MDL No.: MFCD00009317
Vzorec:
C₇H₁₂O₃ MW: 144,17 g/mol Bod varu: 190…192 °C Hustota: 1,012 Bod vzplanutí: 77 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009317 CAS číslo: 4949-44-4 EINECS: 225-593-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...