Ammonium nickel(II) sulphate hexahydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ammonium nickel sulphate hexahydrate, Nickel ammonium sulphate Ammonium disulphatonickelate, Nickel ammonium sulphate hexahydrate
A1827-100G
A1827-500G
SIALA1827-100GEA
1890
CZK
SIALA1827-100G
SIALA1827-500G
Ammonium nickel(II) sulphate hexahydrate, Sigma-Aldrich®
Ammonium nickel (II) sulphate hexahydrate
Vzorec:
H₈N₂NiO₈S₂·6H₂O MW: 394,99 g/mol |
MDL Number:
MFCD00150260 CAS číslo: 7785-20-8 EINECS: 239-793-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...