Tisk…

Ammonium nickel(II) sulphate hexahydrate

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Ammonium nickel sulphate hexahydrate, Nickel ammonium sulphate Ammonium disulphatonickelate, Nickel ammonium sulphate hexahydrate

12519.22 12519.36
12519.22EA 1600 CZK
12519.22 12519.36
Ammonium nickel(II) sulphate hexahydrate
Ammonium nickel(II) sulphate hexahydrate

CAS No.: 7785-20-8 Molecular Formula: H8N2NiO8S2.6H2O Formula Weight: 394.99 (286.90anhy) Physical Form: Large Crystals Density: 1.923 Appearance: Green Odour: Odourless Merck Reference: 14,6501 UN No.: 3077 MDL No.: MFCD00150260

Vzorec: H₈N₂NiO₈S₂·6H₂O
Hustota: 1,923
Storage Temperature: Ambient
MDL Number: MFCD00150260
CAS číslo: 7785-20-8
EINECS: 239-793-5
ADR: 9,III
Merck Index: 14,6501

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR