Aluminium tri-sec-butanolate ≥95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide
Packaged under argon in resealable ChemSeal™ bottles.
43712.22
43712.36
43712.22EA
1300
CZK
43712.22
43712.36
Aluminium tri-sec-butanolate ≥95%
Aluminium tri-sec-butanolate
Packaged under Argon in resealable ChemSeal® bottles
Purity: 95%
CAS No.: 2269-22-9
Molecular Formula: Al[OCH(CH3)CH2CH3]3
Formula Weight: 246.32
Flash Point: 27°(82°F)
Physical Form: Liquid
Density: 0.967
Refractive Index: 1.4390
Sensitivity: Moisture Sensitive
Odour: Alcohol-like
Boiling Point: 200-206°/30mm
Beilstein Reference: 3910908
UN No.: 1992
MDL No.: MFCD00009327
Vzorec:
C₁₂H₂₇AlO₃ MW: 246,33 g/mol Bod varu: 200…206 °C (30 mmHg) Hustota: 0,967 Bod vzplanutí: 27 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009327 CAS číslo: 2269-22-9 EINECS: 218-871-2 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...