Tisk…

Aluminium tributanolate ≥90%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Aluminium n-butoxide

87754.09 87754.18
87754.09EA 4290 CZK
87754.09 87754.18
Aluminium tributanolate ≥90%
Aluminium tributanolate

Purity: 90+% CAS No.: 3085-30-1 Molecular Formula: C12H27AlO3 Formula Weight: 246.32 Physical Form: Lump Sensitivity: Moisture Sensitive UN No.: 3181 MDL No.: MFCD00015237

Vzorec: C₁₂H₂₇AlO₃
MW: 246.33
Storage Temperature: Ambient
MDL Number: MFCD00015237
CAS číslo: 3085-30-1
EINECS: 221-394-2
UN: 3181
ADR: 4.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR