Tisk…

1-Chloroadamantane ≥98%

Dodavatel: Thermo Fisher Scientific

15169.06 15169.14 15169.22
15169.06EA 930 CZK
15169.06 15169.14 15169.22
1-Chloroadamantane ≥98%
1-Chloroadamantane

Purity: 98% CAS No.: 935-56-8 Molecular Formula: C10H15Cl Formula Weight: 170.68 Physical Form: Crystalline Melting Point: 165° MDL No.: MFCD00075627

Vzorec: C₁₀H₁₅Cl
MW: 170.68
Bod tání: 165 °C
Hustota: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00075627
CAS číslo: 935-56-8
EINECS: 213-305-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR