Tisk…

1-Bromoadamantane ≥99%

Dodavatel: Thermo Fisher Scientific

A14893.14 A14893.22
A14893.14EA 1770 CZK
A14893.14 A14893.22
1-Bromoadamantane ≥99%
1-Bromoadamantane

Purity: 99% CAS No.: 768-90-1 Molecular Formula: C10H15Br Formula Weight: 215.14 Melting Point: 117-121° Beilstein Reference: 1098857

Vzorec: C₁₀H₁₅Br
Bod varu: 226 °C (1013 hPa)
Bod tání: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00074721
CAS číslo: 768-90-1
EINECS: 212-200-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR