Tisk…

2,5-Dimethylthiophene ≥98%

Dodavatel: Thermo Scientific
Varování

A10869.09 A10869.14
A10869.09EA 1510 CZK
A10869.09 A10869.14
2,5-Dimethylthiophene ≥98%
2,5-Dimethylthiophene

Purity: 98+% CAS No.: 638-02-8 Molecular Formula: C6H8S Formula Weight: 112.19 Flash Point: 24°(75°F) Refractive Index: 1.5120 Odour: Characteristic Melting Point: -63° Boiling Point: 134-136° Beilstein Reference: 106450 UN No.: 1993 MDL No.: MFCD00005452

Vzorec: C₆H₈S
MW: 112,2 g/mol
Bod varu: 134…136 °C
Bod tání: –63 °C
Hustota: 0,984
Bod vzplanutí: 24 °C (75 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005452
CAS číslo: 638-02-8
EINECS: 211-313-9
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR