Tisk…

Tetrabromothiophene ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A13009.14 A13009.22
A13009.14EA 3540 CZK
A13009.14 A13009.22
Tetrabromothiophene ≥99%
Tetrabromothiophene

Purity: 99% CAS No.: 3958-03-0 Molecular Formula: C4Br4S Formula Weight: 399.72 Odour: Odourless Melting Point: 113-117° Boiling Point: 324-327° Beilstein Reference: 383664 MDL No.: MFCD00005419

Vzorec: C₄Br₄S
Bod varu: 324…327 °C
Bod tání: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00005419
CAS číslo: 3958-03-0
EINECS: 223-554-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR