3-Bromothiophene ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A14022.18
A14022.09
A14022.30
A14022.18EA
4120
CZK
A14022.18
A14022.09
A14022.30
3-Bromothiophene ≥97%
3-Bromothiophene
Purity: 97%
CAS No.: 872-31-1
Molecular Formula: C4H3BrS
Formula Weight: 163.04
Flash Point: 63°(145°F)
Refractive Index: 1.5910
Odour: Pungent
Boiling Point: 157-158°
Beilstein Reference: 105338
UN No.: 2810
MDL No.: MFCD00005464
Vzorec:
C₄H₃BrS Bod varu: 157…158 °C Bod tání: –10 °C Hustota: 1,722 Bod vzplanutí: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005464 CAS číslo: 872-31-1 EINECS: 212-821-3 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...