2,3-Dibromothiophene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A12998.06
A12998.14
A12998.06EA
3240
CZK
A12998.06
A12998.14
2,3-Dibromothiophene ≥98%
2,3-Dibromothiophene
Purity: 98+%
CAS No.: 3140-93-0
Molecular Formula: C4H2Br2S
Formula Weight: 241.94
Flash Point:
Vzorec:
C₄H₂Br₂S Bod varu: 215…216 °C Hustota: 2,135 Bod vzplanutí: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005418 CAS číslo: 3140-93-0 EINECS: 221-542-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...