Tisk…

2,3,5-Collidine ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2,3,5-Trimethylpyridine

B23537.06 B23537.14 B23537.22
B23537.06EA 1030 CZK
B23537.06 B23537.14 B23537.22
2,3,5-Collidine ≥99%
2,3,5-Collidine

Purity: 99% CAS No.: 695-98-7 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 74°(165°F) Refractive Index: 1.5075 Odour: Unpleasant Boiling Point: 182-184° Beilstein Reference: 108832 UN No.: 2810 MDL No.: MFCD00126840

Vzorec: C₈H₁₁N
MW: 121,18 g/mol
Bod varu: 182…184 °C
Hustota: 0,935
Bod vzplanutí: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00126840
CAS číslo: 695-98-7
EINECS: 211-786-1
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR