3-Pikolin ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A14012.0F
A14012.AE
A14012.0FEA
4740
CZK
A14012.0F
A14012.AE
3-Pikolin ≥99%
3-Pikolin
Purity: 99%
CAS No.: 108-99-6
Molecular Formula: C6H7N
Formula Weight: 93.13
Flash Point: 36°(96°F)
Refractive Index: 1.5055
Melting Point: -18°
Boiling Point: 143-144°
Beilstein Reference: 1366
Merck Reference: 14,7401
UN No.: 2313
MDL No.: MFCD00006402
Vzorec:
C₆H₇N Bod varu: 143…144 °C Bod tání: –18 °C Hustota: 0,955 Bod vzplanutí: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006402 CAS číslo: 108-99-6 EINECS: 203-636-9 UN: 2313 ADR: 3,III Merck Index: 14,7401 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...