4-Methoxypyridine ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
L19625.06
L19625.14
L19625.06EA
3170
CZK
L19625.06
L19625.14
4-Methoxypyridine ≥98%
4-Methoxypyridine
Purity: 98+%
CAS No.: 620-08-6
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 74°(165°F)
Refractive Index: 1.5210
Melting Point: 4°
Boiling Point: 191°
Beilstein Reference: 108211
MDL No.: MFCD00674049
Vzorec:
C₆H₇NO MW: 109,13 g/mol Bod varu: 191 °C Bod tání: 4 °C Hustota: 1,075 Bod vzplanutí: 74 °C (165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00674049 CAS číslo: 620-08-6 EINECS: 210-624-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...