2-Aminopyridine 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Pyridylamine, Pyridin-2-amine
A12374.0B
A12374.14
A12374.30
A12374.0BEA
6900
CZK
A12374.0B
A12374.14
A12374.30
2-Aminopyridine 99%
2-Aminopyridine
Purity: 99%
CAS No.: 504-29-0
Molecular Formula: C5H6N2
Formula Weight: 94.12
Flash Point: 92°(197°F)
Melting Point: 56-60°
Boiling Point: 209-211°
Beilstein Reference: 105785
Merck Reference: 14,473
UN No.: 2671
MDL No.: MFCD00006312
Vzorec:
C₅H₆N₂ Bod varu: 209…211 °C Bod tání: 56…60 °C Hustota: 1,24 g/cm³ (20 °C) Bod vzplanutí: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006312 CAS číslo: 504-29-0 EINECS: 207-988-4 UN: 2671 ADR: 6.1,II Merck Index: 14,473 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...