Tisk…

Pentafluoropyridine ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2,3,4,5,6-Perfluoropyridine, 2,3,4,5,6-Pentafluoropyridine

A19656.06 A19656.14
A19656.06EA 975 CZK
A19656.06 A19656.14
Pentafluoropyridine ≥99%
Pentafluoropyridine

Purity: 99% CAS No.: 700-16-3 Molecular Formula: C5F5N Formula Weight: 169.05 Flash Point: 23°(73°F) Refractive Index: 1.3870 Boiling Point: 83-85° Beilstein Reference: 1427064 UN No.: 2924 MDL No.: MFCD00006225

Vzorec: C₅F₅N
MW: 169.05
Bod varu: 83…85 °C
Bod tání: –41 °C
Hustota: 1.618
Bod vzplanutí: 23 °C (73 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006225
CAS číslo: 700-16-3
EINECS: 211-839-9
UN: 2924
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR