Tisk…

2-Chloro-6-methoxypyridine ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L05997.14 L05997.06
L05997.14EA 3420 CZK
L05997.14 L05997.06
2-Chloro-6-methoxypyridine ≥98%
2-Chloro-6-methoxypyridine

Purity: 98% CAS No.: 17228-64-7 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Flash Point: 76°(168°F) Refractive Index: 1.5280 Boiling Point: 185-186° Beilstein Reference: 1364500 MDL No.: MFCD00006265

Vzorec: C₆H₆ClNO
Bod varu: 185…186 °C
Hustota: 1,207
Bod vzplanutí: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006265
CAS číslo: 17228-64-7
EINECS: 241-264-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR