3-Chloropyridine 99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
110020250
110021000
ACRO110020250EA
2220
CZK
ACRO110020250
ACRO110021000
3-Chloropyridine 99%
3-Chloropyridine
Vzorec:
C₅H₄ClN Bod varu: 147…150 °C (760 mmHg) Bod tání: -0,5 °C Hustota: 1,19 g/cm³ Bod vzplanutí: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006375 CAS číslo: 626-60-8 EINECS: 210-955-7 ADR: 3,I |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5310 to 1.5340 (20°C, 589 nm) |
GC | ≥98.5 % |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Substance darkened by H2SO4 | Passes test |
Carbonyl compounds | ≤0.001 % (each of acetone, formaldehyde, acetaldehyde) |
Titrable acid | ≤0.0003 meq/g |
Titrable base | ≤0.0002 meq/g |
KMnO4-reducing substance | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...