2,5-Dichloropyridine ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A15442.14
A15442.18
A15442.14EA
3150
CZK
A15442.14
A15442.18
2,5-Dichloropyridine ≥98%
2,5-Dichloropyridine
Purity: 98%
CAS No.: 16110-09-1
Molecular Formula: C5H3Cl2N
Formula Weight: 147.99
Flash Point: 112°(233°F)
Odour: Unpleasant
Boiling Point: 190-191°
Beilstein Reference: 108886
UN No.: 2811
MDL No.: MFCD00006239
Vzorec:
C₅H₃Cl₂N MW: 147,99 g/mol Bod varu: 190…191 °C Bod tání: 59…62 °C Bod vzplanutí: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006239 CAS číslo: 16110-09-1 EINECS: 240-278-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...