5-Bromo-2-iodopyridine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L19610.14
L19610.06
L19610.14EA
6220
CZK
L19610.14
L19610.06
5-Bromo-2-iodopyridine ≥98%
5-Bromo-2-iodopyridine
Purity: 98%
CAS No.: 223463-13-6
Molecular Formula: C5H3BrIN
Formula Weight: 283.89
Melting Point: 113-116°
Beilstein Reference: 2119
Vzorec:
C₅H₃BrIN Bod tání: 113…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00691506 CAS číslo: 223463-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...