Tisk…

5-Bromo-2-chloropyridine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B24862.06 B24862.14
B24862.06EA 1130 CZK
B24862.06 B24862.14
5-Bromo-2-chloropyridine ≥98%
5-Bromo-2-chloropyridine

Purity: 98% CAS No.: 53939-30-3 Molecular Formula: C5H3BrClN Formula Weight: 192.44 Melting Point: 68-72° Beilstein Reference: 108887

Vzorec: C₅H₃BrClN
MW: 192.44
Bod varu: 205
Bod tání: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD01318951
CAS číslo: 53939-30-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR