3-Bromopyridine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A12894.14
A12894.22
A12894.36
A12894.14EA
1950
CZK
A12894.14
A12894.22
A12894.36
3-Bromopyridine ≥98%
3-Bromopyridine
Purity: 98+%
CAS No.: 626-55-1
Molecular Formula: C5H4BrN
Formula Weight: 158.00
Flash Point: 83°(181°F)
Refractive Index: 1.5700
Odour: Characteristic
Melting Point: -27°
Boiling Point: 172-174°
Beilstein Reference: 105880
UN No.: 2810
MDL No.: MFCD00006373
Vzorec:
C₅H₄BrN Bod varu: 172…174 °C Bod tání: –27 °C Hustota: 1,617 Bod vzplanutí: 72 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006373 CAS číslo: 626-55-1 EINECS: 210-952-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...