2,5-Dibromopyridine ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A10937.18
A10937.09
A10937.18EA
5740
CZK
A10937.18
A10937.09
2,5-Dibromopyridine ≥97%
2,5-Dibromopyridine
Purity: 97%
CAS No.: 624-28-2
Molecular Formula: C5H3Br2N
Formula Weight: 236.90
Melting Point: 92-96°
Boiling Point: 249°
MDL No.: MFCD00006221
Vzorec:
C₅H₃Br₂N Bod varu: 249 °C Bod tání: 92…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006221 CAS číslo: 624-28-2 EINECS: 210-839-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...