Tetrahydrofuran ≥99.7% unstabilised for HPLC, for UV spectrophotometry
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Butylene oxide, THF, Diethylene oxide
22904.AK
22904.K2
22904.0F
22904.AKEA
1690
CZK
22904.AK
22904.K2
22904.0F
Tetrahydrofuran ≥99.7% unstabilised for HPLC, for UV spectrophotometry
Tetrahydrofuran
Purity: 99.7+% min
CAS No.: 109-99-9
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.889
Refractive Index: 1.4070
Appearance: Colourless
Odour: Ether-like
Melting Point: -108°
Boiling Point: 66°
Beilstein Reference: 102391
Merck Reference: 14,9211
UN No.: 2056
MDL No.: MFCD00005356
Vzorec:
C₄H₈O MW: 72.11 Bod varu: 66 °C Bod tání: –108 °C Hustota: 0.889 Bod vzplanutí: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005356 CAS číslo: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3 II REACH: 05-2114615188-48 Merck Index: 14,9211 |