Naftalen ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A13188.0B
A13188.0I
A13188.30
A13188.0BEA
2480
CZK
A13188.0B
A13188.0I
A13188.30
Naftalen ≥99%
Naftalen
Purity: 99+%
CAS No.: 91-20-3
Molecular Formula: C10H8
Formula Weight: 128.17
Flash Point: 78°(172°F)
Refractive Index: 1.5821
Odour: Aromatic
Melting Point: 80-82°
Boiling Point: 218°
Beilstein Reference: 1421310
Merck Reference: 14,6370
UN No.: 1334
MDL No.: MFCD00001742
Vzorec:
C₁₀H₈ Bod varu: 218 °C Bod tání: 80…82 °C Hustota: 0,963 Bod vzplanutí: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001742 CAS číslo: 91-20-3 EINECS: 202-049-5 UN: 1334 ADR: 4.1,III Merck Index: 14,6370 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...