Tisk…

1-Naphthaldehyde ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: α-Naphthaldehyde

A15610.0B A15610.18 A15610.30
A15610.0BEA 16990 CZK
A15610.0B A15610.18 A15610.30
1-Naphthaldehyde ≥97%
1-Naphthaldehyde

Purity: 97% CAS No.: 66-77-3 Molecular Formula: C11H8O Formula Weight: 156.18 Flash Point: 112°(233°F) Refractive Index: 1.6530 Odour: Pungent Melting Point: 1-2° Boiling Point: 160-161°/15mm Beilstein Reference: 386082 MDL No.: MFCD00004003

Vzorec: C₁₁H₈O
MW: 156,18 g/mol
Bod varu: 160…161 °C (15 mmHg)
Bod tání: 1…2 °C
Hustota: 1,148
Bod vzplanutí: 112 °C (233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004003
CAS číslo: 66-77-3
EINECS: 200-633-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR