Tisk…

Rhenium pentachloride ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Rhenium(V) chloride

43294.09 43294.04
43294.09EA 48590 CZK
43294.09 43294.04
Rhenium pentachloride ≥99.9% (metals basis)
Rhenium pentachloride

Purity: 99.9% CAS No.: 13596-35-5 Molecular Formula: ReCl5 Formula Weight: 363.47 Physical Form: -50 Mesh Powder Sensitivity: Moisture Sensitive MDL No.: MFCD00011197

Vzorec: Cl₅Re
MW: 363.47
Bod tání: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00011197
CAS číslo: 13596-35-5
EINECS: 237-042-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR