Tisk…

Chromium(III) telluride ≥99.5% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

40206.06 40206.14
40206.06EA 2230 CZK
40206.06 40206.14
Chromium(III) telluride ≥99.5% (metals basis)
Chromium(III) telluride

Purity: 99.5% CAS No.: 12053-39-3 Molecular Formula: Cr2Te3 Formula Weight: 486.79 Physical Form: -60+100 Mesh Powder Density: 7.0 Appearance: Gray-black Odour: Odourless Melting Point: ca 1300° MDL No.: MFCD00016040

Vzorec: Cr₂Te₃
MW: 486.79
Bod tání: ∼1300 °C
Hustota: 7.0
Storage Temperature: Ambient
MDL Number: MFCD00016040
CAS číslo: 12053-39-3
EINECS: 235-003-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR