Molybdenum diselenide ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
13112.06
13112.14
13112.06EA
3210
CZK
13112.06
13112.14
Molybdenum diselenide ≥99.9% (metals basis)
Molybdenum diselenide
Purity: 99.9%
CAS No.: 12058-18-3
Molecular Formula: MoSe2
Formula Weight: 253.87
Physical Form: -325 Mesh Powder
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00014216
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...