Tisk…

Oxid chromový ≥99.9% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10691.14 10691.22 10691.36
10691.14EA 2760 CZK
10691.14 10691.22 10691.36
Oxid chromový ≥99.9% (metals basis), Puratronic®
Oxid chromový

Purity: 99.9% CAS No.: 1333-82-0 Molecular Formula: CrO3 Formula Weight: 99.99 Physical Form: Crystalline Density: 2.7 Appearance: Dark red Melting Point: 196° Boiling Point: 250° dec. Merck Reference: 14,2235 UN No.: 1463 MDL No.: MFCD00010952

Vzorec: CrO₃
Bod varu: 250 °C (rozkl.)
Bod tání: 196 °C
Hustota: 2,7
Storage Temperature: Ambient
MDL Number: MFCD00010952
CAS číslo: 1333-82-0
EINECS: 215-607-8
UN: 1463
ADR: 5.1,II
REACH: 01-2119458868-17
Merck Index: 14,2235

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR