Tisk…

Zinc(II) telluride ≥99.998% (metals basis)

Dodavatel: Thermo Scientific
Varování

36595.09 36595.18
36595.09EA 4640 CZK
36595.09 36595.18
Zinc(II) telluride ≥99.998% (metals basis)
Zinc(II) telluride

Purity: 99.998% CAS No.: 1315-11-3 Molecular Formula: ZnTe Formula Weight: 192.97 Physical Form: 3-12mm Lump Density: 6.34 Refractive Index: 3.56 Appearance: Red Odour: Odourless Melting Point: 1240° Merck Reference: 14,10162 MDL No.: MFCD00043079

Vzorec: TeZn
MW: 192,99 g/mol
Bod tání: 1240 °C
Hustota: 6,34
Storage Temperature: Ambient
MDL Number: MFCD00043079
CAS číslo: 1315-11-3
EINECS: 215-260-2
Merck Index: 14,10162

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR