Tisk…

Mercury(II) telluride ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

36666.04 36666.09
36666.04EA 4600 CZK
36666.04 36666.09
Mercury(II) telluride ≥99.999% (metals basis)
Mercury(II) telluride

Purity: 99.999% CAS No.: 12068-90-5 Molecular Formula: HgTe Formula Weight: 328.19 Physical Form: 3-12mm Fused Lump Density: 8.63 Appearance: Gray to black Odour: Odourless Melting Point: 673° UN No.: 2025 MDL No.: MFCD00016133

Vzorec: HgTe
Bod tání: 673 °C
Hustota: 8,63
Storage Temperature: Ambient
MDL Number: MFCD00016133
CAS číslo: 12068-90-5
EINECS: 235-108-9
UN: 2025
ADR: 6.1,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR