Cadmium(II) telluride ≥99.999% (metals basis)
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Cadmium telluride
14365.14
14365.22
14365.14EA
6280
CZK
14365.14
14365.22
Cadmium(II) telluride ≥99.999% (metals basis)
Cadmium(II) telluride
Purity: 99.999%
CAS No.: 1306-25-8
Molecular Formula: CdTe
Formula Weight: 240.00
Physical Form: Polycrystalline Lump
Density: 6.20
Appearance: Black
Odour: Odourless
Melting Point: 1041°
Merck Reference: 14,1629
UN No.: 3077
MDL No.: MFCD00015998
Vzorec:
CdTe MW: 240,01 g/mol Bod tání: 1041 °C Hustota: 6,20 Storage Temperature: Ambient |
MDL Number:
MFCD00015998 CAS číslo: 1306-25-8 EINECS: 215-149-9 UN: 3077 ADR: 9,III Merck Index: 14,1629 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...