Tisk…

Oxid zinečnatý ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Zinci oxidum, Zinc (II) oxide

11137.09 11137.18 11137.30
11137.09EA 2320 CZK
11137.09 11137.18 11137.30
Oxid zinečnatý ≥99.999% (metals basis), Puratronic®
Zinc oxide

Purity: 99.999% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: Powder Density: 5.606 Melting Point: 1975° Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300

Vzorec: ZnO
Bod tání: 1975 °C
Hustota: 5,606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS číslo: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR