Tisk…

Oxid stříbrný ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: di-Silver oxide, Disilver oxide, Silver oxide

42577.03 42577.06 42577.14 42577.49
42577.03EA 1090 CZK
42577.03 42577.06 42577.14 42577.49
Oxid stříbrný ≥99.99% (metals basis)
Oxid stříbrný

Purity: 99.99% CAS No.: 20667-12-3 Molecular Formula: Ag2O Formula Weight: 231.74 Physical Form: Powder Density: 7.2 Appearance: Dark brown Odour: Odourless Melting Point: 230° dec. Merck Reference: 14,8521 UN No.: 1479 MDL No.: MFCD00003404

Vzorec: Ag₂O
MW: 231.74
Bod tání: 230 °C (rozkl.)
Hustota: 7.2
Storage Temperature: Ambient
MDL Number: MFCD00003404
CAS číslo: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1 II
Merck Index: 14,8521

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR